Assay Details
Target Gene Details
Entrez Gene ID: | 54511 |
Gene Name: | 3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1 |
Gene Aliases: |
bA418P12.1, er-cHL |
Location: |
Chr.6:55434373-55579214 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGCLL1 | NM_001042406.1 | 9 | 1897 | NP_001035865.1 |
| NM_001287741.1 | 8 | 1801 | NP_001274670.1 | |
| NM_001287746.1 | 10 | 1993 | NP_001274675.1 | |
| NM_001287753.1 | 6 | 1588 | NP_001274682.1 | |
| NM_019036.2 | 10 | 1987 | NP_061909.2 | |
| NR_109869.1 | 5 | 1399 | ||
| AF131827.1 | 5 | 1291 | AAD20054.1 | |
| AK055075.1 | 10 | 1878 | ||
| AK127587.1 | 10 | 1958 | BAC87045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018622 | Chr.6:55433056 - 55455513 on Build GRCh38 | Loss |
|
| dgv5965n100 | Chr.6:55372925 - 55522683 on Build GRCh38 | Gain |
|
| nsv470819 | Chr.6:55372139 - 55504017 on Build GRCh38 | Gain |
|
| nsv5311 | Chr.6:55394592 - 55439247 on Build GRCh38 | Deletion |
|
| nsv1034037 | Chr.6:55357611 - 56204802 on Build GRCh38 | Gain |
|
| nsv462948 | Chr.6:55373293 - 55504017 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map