Assay Details
Target Gene Details
Entrez Gene ID: | 84694 |
Gene Name: | gap junction protein alpha 10 |
Gene Aliases: |
CX62 |
Location: |
Chr.6:89894469-89896100 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA10 | NM_032602.1 | 1 | 607 | NP_115991.1 |
| AF296766.1 | 1 | 607 | AAK51676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604125 | Chr.6:89751706 - 90189842 on Build GRCh38 | Gain |
|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
| esv3609910 | Chr.6:89880424 - 89900299 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map