Assay Details
Target Gene Details
Entrez Gene ID: | 4225 |
Gene Name: | meprin A subunit beta |
Gene Aliases: |
- |
Location: |
Chr.18:32190024-32222607 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEP1B | NM_001308171.1 | NP_001295100.1 | ||
| NM_005925.2 | NP_005916.2 | |||
| XM_011526013.2 | XP_011524315.1 | |||
| XM_011526014.2 | XP_011524316.1 | |||
| AY695931.1 | AAU05377.1 | |||
| BC136559.1 | ||||
| BC144244.1 | ||||
| X81333.1 | CAA57107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2249 | Chr.18:32160231 - 32192685 on Build GRCh38 | Insertion |
|
| nsv1056007 | Chr.18:32132011 - 32319601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map