Assay Details
Target Gene Details
Entrez Gene ID: | 23274 |
Gene Name: | C-type lectin domain family 16 member A |
Gene Aliases: |
Gop-1, KIAA0350 |
Location: |
Chr.16:10944488-11193272 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC16A | NM_001243403.1 | NP_001230332.1 | ||
| NM_015226.2 | NP_056041.1 | |||
| XM_005255210.1 | XP_005255267.1 | |||
| XM_005255211.1 | XP_005255268.1 | |||
| XM_005255213.1 | XP_005255270.1 | |||
| XM_005255214.1 | XP_005255271.1 | |||
| XM_005255215.3 | XP_005255272.1 | |||
| XM_005255216.1 | XP_005255273.1 | |||
| XM_006720870.3 | XP_006720933.1 | |||
| XM_011522434.1 | XP_011520736.1 | |||
| XM_011522435.1 | XP_011520737.1 | |||
| XM_011522436.2 | XP_011520738.1 | |||
| XM_011522437.2 | XP_011520739.1 | |||
| XM_011522438.2 | XP_011520740.1 | |||
| XM_011522439.2 | XP_011520741.1 | |||
| XM_011522440.2 | XP_011520742.1 | |||
| XM_017023089.1 | XP_016878578.1 | |||
| AB002348.3 | ||||
| BC112897.1 | AAI12898.1 | |||
| DB069341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map