Assay Details
Target Gene Details
Entrez Gene ID: | 3321 |
Gene Name: | immunoglobulin superfamily member 3 |
Gene Aliases: |
EWI-3, LCDD, V8 |
Location: |
Chr.1:116574398-116667755 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF3 | NM_001007237.2 | NP_001007238.1 | ||
| NM_001542.3 | NP_001533.2 | |||
| XM_005270793.2 | XP_005270850.1 | |||
| XM_005270794.4 | XP_005270851.1 | |||
| XM_006710593.3 | XP_006710656.1 | |||
| XM_011541315.1 | XP_011539617.1 | |||
| XM_011541316.2 | XP_011539618.1 | |||
| AB007935.2 | ||||
| AI968231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482103 | Chr.1:116574409 - 116667692 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs551130466] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map