Assay Details
Target Gene Details
Entrez Gene ID: | 140691 |
Gene Name: | tripartite motif containing 69 |
Gene Aliases: |
HSD-34, HSD34, RNF36, Trif |
Location: |
Chr.15:44736362-44767829 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM69 | NM_001301144.1 | 1 | 264 | NP_001288073.1 |
| NM_001301145.1 | 1 | 264 | NP_001288074.1 | |
| NM_001301146.1 | 1 | 264 | NP_001288075.1 | |
| NM_080745.4 | 1 | 264 | NP_542783.2 | |
| NM_182985.4 | 1 | 264 | NP_892030.3 | |
| AF302088.1 | 1 | 112 | AAL55810.1 | |
| AF302089.1 | 1 | 112 | AAL55811.1 | |
| AK292252.1 | 1 | 108 | ||
| AL360161.1 | 1 | 99 | ||
| AY305385.1 | AAQ75551.1 | |||
| BC024199.1 | 1 | 82 | AAH24199.1 | |
| BC033314.1 | 1 | 60 | AAH33314.1 | |
| BC047945.1 | 1 | 101 | AAH47945.1 | |
| BI465168.1 | 1 | 96 | ||
| BI827618.1 | 1 | 264 | ||
| BI829233.1 | 1 | 82 | ||
| BM559467.1 | 1 | 110 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832990 | Chr.15:44698567 - 44870516 on Build GRCh38 | Gain |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map