Assay Details
Target Gene Details
Entrez Gene ID: | 93978 |
Gene Name: | C-type lectin domain family 6 member A |
Gene Aliases: |
CLEC4N, CLECSF10 |
Location: |
Chr.12:8455995-8478330 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC6A | NM_001007033.1 | NP_001007034.1 | ||
| NM_001317999.1 | NP_001304928.1 | |||
| AK308011.1 | ||||
| AY321309.1 | AAQ83725.1 | |||
| AY365135.1 | AAR13071.1 | |||
| BC132933.1 | ||||
| BC132935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv1035270 | Chr.12:8466966 - 8521466 on Build GRCh38 | Gain |
|
| nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss |
|
| nsv518125 | Chr.12:8463213 - 8518678 on Build GRCh38 | Loss |
|
| dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map