Assay Details
Target Gene Details
Entrez Gene ID: | 2553 |
Gene Name: | GA binding protein transcription factor beta subunit 1 |
Gene Aliases: |
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2 |
Location: |
Chr.15:50277192-50355429 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABPB1 | NM_001320910.1 | 10 | 1741 | NP_001307839.1 |
| NM_001320915.1 | 10 | 1738 | NP_001307844.1 | |
| NM_005254.5 | 9 | 1696 | NP_005245.2 | |
| NM_016654.4 | 9 | 1660 | NP_057738.1 | |
| XM_005254274.3 | 9 | 1404 | XP_005254331.1 | |
| XM_011521426.2 | 9 | 1657 | XP_011519728.1 | |
| XM_017022053.1 | 10 | 1439 | XP_016877542.1 | |
| XM_017022054.1 | 10 | 1397 | XP_016877543.1 | |
| AK301350.1 | 9 | 1486 | ||
| AK303901.1 | 8 | 1304 | ||
| BC036080.1 | 9 | 1454 | AAH36080.1 | |
| BC050702.1 | 9 | 1401 | AAH50702.1 | |
| D13317.1 | 9 | 1441 | BAA02574.1 | |
| EU159454.1 | 8 | 1274 | ||
| U13045.1 | 9 | 1441 | AAA65707.1 | |
| U13046.1 | 9 | 1405 | AAA65708.1 |
More Information
Set Membership: |
|

Genomic Map