Assay Details
Target Gene Details
Entrez Gene ID: | 10242 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 2 |
Gene Aliases: |
- |
Location: |
Chr.3:178536298-178844429 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB2 | NM_001278911.1 | 1 | 182 | NP_001265840.1 |
| NM_005832.4 | 1 | 182 | NP_005823.1 | |
| NM_181361.2 | NP_852006.1 | |||
| XM_011512325.2 | XP_011510627.1 | |||
| AA904191.1 | ||||
| AF209747.1 | 1 | 182 | AAF36562.1 | |
| AK298291.1 | ||||
| BC017825.1 | AAH17825.1 | |||
| BG701449.1 | ||||
| DA532966.1 |
Target Gene Details
Entrez Gene ID: | 104797538 |
Gene Name: | KCNMB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:178525467-178860405 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB2-AS1 | NR_126560.1 | |||
| NR_126561.1 | ||||
| AF279780.1 | ||||
| AY769439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524447 | Chr.3:178539801 - 178638481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map