Assay Details
Target Gene Details
Entrez Gene ID: | 10291 |
Gene Name: | splicing factor 3a subunit 1 |
Gene Aliases: |
PRP21, PRPF21, SAP114, SF3A120 |
Location: |
Chr.22:30331988-30356947 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3A1 | NM_005877.5 | 16 | 2816 | NP_005868.1 |
| AK098463.1 | 9 | 1582 | ||
| AK303273.1 | 17 | 2741 | ||
| BC001976.1 | 16 | 2756 | AAH01976.1 | |
| BC007684.2 | 16 | 2783 | AAH07684.1 | |
| BQ447452.1 | 1 | 621 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834174 | Chr.22:30223771 - 30350926 on Build GRCh38 | Gain+Loss |
|
| nsv1064350 | Chr.22:30328159 - 30346136 on Build GRCh38 | Gain |
|
| nsv3593 | Chr.22:30309991 - 30355133 on Build GRCh38 | Deletion |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map