Assay Details
Target Gene Details
Entrez Gene ID: | 1828 |
Gene Name: | desmoglein 1 |
Gene Aliases: |
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1 |
Location: |
Chr.18:31318089-31359246 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1 | NM_001942.3 | 11 | 1777 | NP_001933.2 |
| AF097935.1 | 11 | 1778 | AAC83817.1 | |
| X56654.1 | 11 | 1642 | CAA39976.1 |
More Information
Set Membership: |
|

Genomic Map