Assay Details
Target Gene Details
Entrez Gene ID: | 23193 |
Gene Name: | glucosidase II alpha subunit |
Gene Aliases: |
G2AN, GIIA, GLUII |
Location: |
Chr.11:62624826-62646726 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GANAB | NM_001278192.1 | 7 | 792 | NP_001265121.1 |
| NM_001278193.1 | 6 | 726 | NP_001265122.1 | |
| NM_001278194.1 | 8 | 959 | NP_001265123.1 | |
| NM_198334.2 | 9 | 1068 | NP_938148.1 | |
| NM_198335.3 | 10 | 1134 | NP_938149.2 | |
| XM_017017412.1 | 7 | 802 | XP_016872901.1 | |
| XM_017017413.1 | 9 | 1184 | XP_016872902.1 | |
| XM_017017414.1 | 9 | 1015 | XP_016872903.1 | |
| AF144074.1 | 10 | 1023 | AAF66685.1 | |
| AJ000332.1 | 9 | 941 | CAA04006.1 | |
| AK026997.1 | 9 | 958 | ||
| AK295806.1 | 6 | 614 | ||
| AK299819.1 | 8 | 842 | ||
| AK302752.1 | 7 | 698 | ||
| AK314370.1 | 9 | 957 | ||
| BC065266.1 | 8 | 835 | AAH65266.1 | |
| D42041.1 | 9 | 939 | BAA07642.1 | |
| EU794613.1 | 9 | 974 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain |
|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map