Assay Details
Target Gene Details
Entrez Gene ID: | 100128496 |
Gene Name: | chromosome 20 open reading frame 78 |
Gene Aliases: |
dJ1068E13.1 |
Location: |
Chr.20:18809351-18830203 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C20orf78 | NM_001242671.2 | NP_001229600.1 | ||
| NM_001302818.1 | NP_001289747.1 | |||
| AW118618.1 | ||||
| BX112808.1 | ||||
| BX334499.2 | ||||
| HY016900.1 | ||||
| HY050710.1 |
Target Gene Details
Entrez Gene ID: | 140856 |
Gene Name: | SCP2 sterol binding domain containing 1 |
Gene Aliases: |
C20orf79, HSD22, dJ1068E13.2 |
Location: |
Chr.20:18813726-18814392 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCP2D1 | NM_178483.2 | 1 | 515 | NP_848578.1 |
| AF418568.1 | 1 | 515 | AAP97287.1 | |
| AF418570.1 | 1 | 515 | AAP97289.1 | |
| AK130970.1 | 1 | 511 | ||
| AY248902.1 | 1 | 472 | AAP20053.1 | |
| BC105121.1 | 1 | 457 | AAI05122.1 | |
| BC112024.1 | 1 | 457 | AAI12025.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833934 | Chr.20:18778246 - 18958077 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map