Assay Details
Target Gene Details
Entrez Gene ID: | 343099 |
Gene Name: | coiled-coil domain containing 18 |
Gene Aliases: |
NY-SAR-41 |
Location: |
Chr.1:93179802-93279037 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC18 | XM_011541361.2 | 1 | 50 | XP_011539663.2 |
| XM_011541372.2 | 1 | 50 | XP_011539674.2 | |
| XM_017001154.1 | 1 | 50 | XP_016856643.1 | |
| XM_017001155.1 | 1 | 50 | XP_016856644.1 | |
| XM_017001156.1 | 1 | 50 | XP_016856645.1 | |
| XM_017001157.1 | 1 | 50 | XP_016856646.1 | |
| XM_017001158.1 | 1 | 50 | XP_016856647.1 | |
| XM_017001159.1 | 1 | 50 | XP_016856648.1 | |
| XM_017001160.1 | 1 | 50 | XP_016856649.1 | |
| XM_017001161.1 | 1 | 50 | XP_016856650.1 | |
| XM_017001162.1 | XP_016856651.1 | |||
| XM_017001164.1 | XP_016856653.1 | |||
| XM_017001165.1 | 1 | 50 | XP_016856654.1 | |
| XM_017001169.1 | 1 | 50 | XP_016856658.1 | |
| AW444474.1 | 1 | 193 |
Target Gene Details
Entrez Gene ID: | 50999 |
Gene Name: | transmembrane p24 trafficking protein 5 |
Gene Aliases: |
CGI-100, p24g2, p28 |
Location: |
Chr.1:93149742-93180689 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED5 | NM_001167830.1 | 1 | 134 | NP_001161302.1 |
| NM_016040.4 | 1 | 134 | NP_057124.3 | |
| NR_030761.1 | 1 | 134 | ||
| AI436590.1 | 1 | 194 | ||
| BC070051.1 | 1 | 134 | AAH70051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map