Assay Details
Target Gene Details
Entrez Gene ID: | 55665 |
Gene Name: | upregulator of cell proliferation |
Gene Aliases: |
URG4 |
Location: |
Chr.7:43875894-43926411 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URGCP | NM_001077663.2 | 6 | 2487 | NP_001071131.1 |
| NM_001077664.2 | 6 | 2259 | NP_001071132.1 | |
| NM_001290075.1 | 6 | 2482 | NP_001277004.1 | |
| NM_001290076.1 | 7 | 2381 | NP_001277005.1 | |
| NM_017920.4 | 5 | 2460 | NP_060390.3 | |
| AB040940.1 | 7 | 2784 | BAA96031.1 | |
| AK000661.1 | 1 | 194 | BAA91312.1 | |
| AK026185.1 | 1 | 1664 | ||
| AK302459.1 | 7 | 2381 | ||
| AK314339.1 | 5 | 2006 | ||
| AL832134.1 | 4 | 4100 | ||
| AL833744.1 | 1 | 588 | CAH56246.1 | |
| AY078404.1 | 5 | 1979 | AAL83710.1 | |
| BC018426.1 | 5 | 1981 | ||
| BX640797.1 | 6 | 2022 | CAE45880.1 | |
| BX648505.1 | 6 | 2027 | ||
| CR749398.1 | 6 | 2039 | CAH18244.1 | |
| DR421788.1 | 1 | 335 |
Target Gene Details
Entrez Gene ID: | 100534592 |
Gene Name: | URGCP-MRPS24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:43866558-43906632 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URGCP-MRPS24 | NM_001204871.1 | NP_001191800.1 | ||
| AI027170.1 | ||||
| FY210844.1 | ||||
| FY212401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map