Assay Details
Target Gene Details
Entrez Gene ID: | 53632 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 3 |
Gene Aliases: |
AMPKG3 |
Location: |
Chr.2:218821853-218831803 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG3 | NM_017431.2 | 10 | 1094 | NP_059127.2 |
| XM_017004343.1 | 10 | 1108 | XP_016859832.1 | |
| AF214519.1 | 8 | 1000 | AAF73987.1 | |
| AJ249977.1 | 10 | 1096 | CAB65117.1 | |
| AK300078.1 | 11 | 1060 | ||
| AK300687.1 | 9 | 1062 | ||
| AK309865.1 | 9 | 1233 | ||
| BC098102.1 | 10 | 1094 | AAH98102.1 | |
| BC098255.1 | 10 | 1094 | AAH98255.1 | |
| BC098277.1 | 10 | 1094 | AAH98277.1 | |
| BC098306.1 | 10 | 1094 | AAH98306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528407 | Chr.2:218822874 - 218833281 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map