Assay Details
Target Gene Details
Entrez Gene ID: | 79607 |
Gene Name: | family with sequence similarity 118 member B |
Gene Aliases: |
- |
Location: |
Chr.11:126211414-126264540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM118B | NM_024556.3 | NP_078832.1 | ||
| XM_005271665.3 | XP_005271722.1 | |||
| XM_011542977.2 | XP_011541279.1 | |||
| XM_011542978.2 | XP_011541280.1 | |||
| XM_017018285.1 | XP_016873774.1 | |||
| XM_017018286.1 | XP_016873775.1 | |||
| AK024756.1 | BAB14987.1 | |||
| AK225081.1 | ||||
| AK315615.1 | ||||
| BC001340.1 | AAH01340.1 | |||
| BC001647.2 | AAH01647.1 | |||
| CR457334.1 | CAG33615.1 |
Target Gene Details
Entrez Gene ID: | 6734 |
Gene Name: | SRP receptor alpha subunit |
Gene Aliases: |
DP, SRPR, Sralpha |
Location: |
Chr.11:126255151-126268982 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRA | XM_017018179.1 | XP_016873668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556529 | Chr.11:126204297 - 126264724 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map