Assay Details
Target Gene Details
Entrez Gene ID: | 9270 |
Gene Name: | integrin subunit beta 1 binding protein 1 |
Gene Aliases: |
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B |
Location: |
Chr.2:9405681-9423569 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB1BP1 | NM_001319066.1 | 7 | 1905 | NP_001305995.1 |
| NM_001319067.1 | 7 | 1807 | NP_001305996.1 | |
| NM_001319068.1 | 7 | 1778 | NP_001305997.1 | |
| NM_001319069.1 | 5 | 1607 | NP_001305998.1 | |
| NM_001319070.1 | 5 | 1577 | NP_001305999.1 | |
| NM_004763.4 | 7 | 1793 | NP_004754.1 | |
| NM_022334.4 | 6 | 1643 | NP_071729.1 | |
| NR_134951.1 | 7 | 1782 | ||
| NR_134952.1 | 6 | 1844 | ||
| NR_134953.1 | 5 | 1536 | ||
| NR_134954.1 | 7 | 1779 | ||
| NR_134955.1 | 7 | 1789 | ||
| NR_134956.1 | 6 | 1714 | ||
| NR_134957.1 | 6 | 1686 | ||
| AF012023.1 | 8 | 1224 | AAB88671.1 | |
| AF012024.1 | 7 | 1074 | AAB88672.1 | |
| CA428076.1 | 1 | 213 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
| esv3414345 | Chr.2:9405772 - 9406570 on Build GRCh38 | Duplication |
|
| esv2478125 | Chr.2:9405537 - 9407116 on Build GRCh38 | Deletion |
|
| dgv6685n54 | Chr.2:9405901 - 9406676 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map