Assay Details
Target Gene Details
Entrez Gene ID: | 200197 |
Gene Name: | TMEM51 antisense RNA 1 |
Gene Aliases: |
C1orf126 |
Location: |
Chr.1:15111815-15152464 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM51-AS1 | NR_027136.1 | |||
| AK074283.1 | BAB85035.1 | |||
| AK124431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833425 | Chr.1:15083330 - 15291728 on Build GRCh38 | Gain |
|
| nsv508913 | Chr.1:15092390 - 15165461 on Build GRCh38 | Insertion |
|
| nsv545591 | Chr.1:14880515 - 15130423 on Build GRCh38 | Loss |
|

Genomic Map