Assay Details
Target Gene Details
Entrez Gene ID: | 25827 |
Gene Name: | F-box and leucine rich repeat protein 2 |
Gene Aliases: |
FBL2, FBL3 |
Location: |
Chr.3:33277422-33422371 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL2 | AK294209.1 | |||
| AK316229.1 |
Target Gene Details
Entrez Gene ID: | 7342 |
Gene Name: | upstream binding protein 1 (LBP-1a) |
Gene Aliases: |
LBP-1B, LBP-1a, LBP1A, LBP1B |
Location: |
Chr.3:33388336-33440405 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBP1 | NM_001128160.1 | NP_001121632.1 | ||
| NM_001128161.1 | NP_001121633.1 | |||
| NM_014517.4 | NP_055332.3 | |||
| AF198487.1 | AAF32274.1 | |||
| AK023274.1 | BAB14501.1 | |||
| AK023278.1 | ||||
| AK293100.1 | ||||
| BC047235.1 | AAH47235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001132 | Chr.3:33386293 - 33565948 on Build GRCh38 | Gain |
|
| esv2759138 | Chr.3:33253615 - 33429981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map