Assay Details
Target Gene Details
Entrez Gene ID: | 344752 |
Gene Name: | arylacetamide deacetylase like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:151733916-151758016 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2 | NM_207365.3 | 1 | 194 | NP_997248.2 |
| BC065724.1 | 1 | 187 | AAH65724.1 | |
| BX647585.1 | 1 | 194 | CAI46075.1 | |
| CU449274.1 |
More Information
Set Membership: |
|

Genomic Map