Assay Details
Target Gene Details
Entrez Gene ID: | 1807 |
Gene Name: | dihydropyrimidinase |
Gene Aliases: |
DHP, DHPase |
Location: |
Chr.8:104379424-104467075 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYS | NM_001385.2 | NP_001376.1 | ||
| XM_005250818.3 | XP_005250875.1 | |||
| XM_006716518.3 | XP_006716581.1 | |||
| AK311261.1 | ||||
| BC034395.1 | AAH34395.1 | |||
| D78011.1 | BAA11189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032775 | Chr.8:104299136 - 104397744 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map