Assay Details
Target Gene Details
Entrez Gene ID: | 7375 |
Gene Name: | ubiquitin specific peptidase 4 |
Gene Aliases: |
UNP, Unph |
Location: |
Chr.3:49277144-49340103 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP4 | NM_003363.3 | NP_003354.2 | ||
| NM_199443.2 | NP_955475.1 | |||
| AF017305.1 | AAC27355.1 | |||
| AF017306.1 | AAC27356.1 | |||
| AK127055.1 | ||||
| AK222725.1 | BAD96445.1 | |||
| AK223292.1 | BAD97012.1 | |||
| AK291795.1 | ||||
| BC125130.1 | ||||
| BC125131.1 | ||||
| U20657.1 | AAB72237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008750 | Chr.3:49171382 - 49324968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map