Assay Details
Target Gene Details
Entrez Gene ID: | 51099 |
Gene Name: | abhydrolase domain containing 5 |
Gene Aliases: |
CDS, CGI58, IECN2, NCIE2 |
Location: |
Chr.3:43690883-43722725 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD5 | NM_016006.4 | 3 | 446 | NP_057090.2 |
| XM_011533779.2 | 3 | 696 | XP_011532081.1 | |
| XM_011533780.1 | 3 | 393 | XP_011532082.1 | |
| AF151816.1 | 3 | 342 | AAD34053.1 | |
| AK313811.1 | 3 | 356 | ||
| BC021958.1 | 3 | 374 | AAH21958.1 | |
| BG721387.1 | 3 | 465 | ||
| KU178625.1 | 3 | 336 | ||
| KU178626.1 | 3 | 336 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834673 | Chr.3:43581493 - 43758384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map