Assay Details
Target Gene Details
Entrez Gene ID: | 79174 |
Gene Name: | cysteine rich with EGF like domains 2 |
Gene Aliases: |
- |
Location: |
Chr.22:49918630-49927540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRELD2 | NM_001135101.2 | NP_001128573.1 | ||
| NM_001284317.1 | NP_001271246.1 | |||
| NM_001284318.1 | NP_001271247.1 | |||
| NM_024324.4 | NP_077300.3 | |||
| NR_104295.1 | ||||
| XM_005261737.2 | XP_005261794.1 | |||
| XM_005261738.4 | XP_005261795.1 | |||
| XM_017028938.1 | XP_016884427.1 | |||
| AJ968414.1 | CAI91316.1 | |||
| AK225930.1 | ||||
| AK308535.1 | ||||
| AK308692.1 | ||||
| AY358355.1 | AAQ88721.1 | |||
| BC002894.1 | AAH02894.1 | |||
| BC050675.1 | AAH50675.1 | |||
| DQ470676.1 | ||||
| DQ470677.1 | ||||
| DQ470678.1 | ||||
| DQ470679.1 | ||||
| DQ470680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1334e212 | Chr.22:49920870 - 49933235 on Build GRCh38 | Loss |
|
| nsv471221 | Chr.22:49880243 - 49927295 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map