Assay Details
Target Gene Details
Entrez Gene ID: | 116228 |
Gene Name: | COX20, cytochrome c oxidase assembly factor |
Gene Aliases: |
FAM36A |
Location: |
Chr.1:244835306-244845063 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX20 | NM_001312871.1 | 5 | 1865 | NP_001299800.1 |
| NM_001312872.1 | 5 | 1873 | NP_001299801.1 | |
| NM_001312873.1 | 3 | 1702 | NP_001299802.1 | |
| NM_001312874.1 | 4 | 1833 | NP_001299803.1 | |
| NM_198076.5 | 4 | 1984 | NP_932342.1 | |
| NR_132419.1 | 3 | 1722 | ||
| NR_132420.1 | 4 | 1817 | ||
| NR_132421.1 | 3 | 1698 | ||
| AK000866.1 | 4 | 1627 | ||
| AK125259.1 | 4 | 1976 | ||
| AK130498.1 | 4 | 1650 | ||
| AK300760.1 | 3 | 1503 |
Target Gene Details
Entrez Gene ID: | 3192 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U |
Location: |
Chr.1:244842123-244864720 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPU | XM_017001115.1 | 15 | 11665 | XP_016856604.1 |
| XM_017001116.1 | 16 | 9389 | XP_016856605.1 | |
| XM_017001117.1 | 15 | 11459 | XP_016856606.1 | |
| AK095297.1 | 2 | 1000 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549493 | Chr.1:244721983 - 244856983 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map