Assay Details
Target Gene Details
Entrez Gene ID: | 150465 |
Gene Name: | tubulin tyrosine ligase |
Gene Aliases: |
- |
Location: |
Chr.2:112482127-112532646 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTL | NM_153712.4 | 7 | 1721 | NP_714923.1 |
| XM_005263599.3 | 8 | 1880 | XP_005263656.1 | |
| XM_011510665.2 | 6 | 1616 | XP_011508967.1 | |
| AB071393.2 | BAC06832.2 | |||
| AK126378.1 | 2 | 940 | ||
| AK126549.1 | 5 | 1213 | ||
| BC036819.1 | 7 | 1733 | AAH36819.1 | |
| BX538316.1 | 3 | 932 | CAD98091.1 | |
| BX647135.1 | 4 | 1019 | ||
| BX649144.1 | 1 | 6091 |
More Information
Set Membership: |
|

Genomic Map