Assay Details
Target Gene Details
Entrez Gene ID: | 84174 |
Gene Name: | Src like adaptor 2 |
Gene Aliases: |
C20orf156, MARS, SLAP-2, SLAP2 |
Location: |
Chr.20:36612519-36646216 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLA2 | NM_032214.3 | NP_115590.1 | ||
| NM_175077.2 | NP_778252.1 | |||
| AF290985.1 | AAL38197.1 | |||
| AF290986.1 | AAL38198.1 | |||
| AF326353.1 | AAL29204.1 | |||
| AK025645.1 | BAB15201.1 | |||
| AK291513.1 | ||||
| BC042041.1 | AAH42041.1 | |||
| BQ052468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map