Assay Details
Target Gene Details
Entrez Gene ID: | 8242 |
Gene Name: | lysine demethylase 5C |
Gene Aliases: |
DXS1272E, JARID1C, MRX13, MRXJ, MRXSCJ, MRXSJ, SMCX, XE169 |
Location: |
Chr.X:53159735-53225422 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM5C | NM_001146702.1 | NP_001140174.1 | ||
| NM_001282622.1 | 26 | 4972 | NP_001269551.1 | |
| NM_004187.3 | 26 | 4975 | NP_004178.2 | |
| XM_005262035.4 | 26 | 4779 | XP_005262092.1 | |
| XM_011530824.2 | 26 | 4710 | XP_011529126.1 | |
| XM_011530825.2 | 25 | 4665 | XP_011529127.1 | |
| XM_011530826.2 | 25 | 4656 | XP_011529128.1 | |
| XM_011530827.2 | XP_011529129.1 | |||
| XM_011530828.1 | XP_011529130.1 | |||
| XM_011530829.2 | 27 | 4857 | XP_011529131.1 | |
| XM_011530830.2 | 27 | 4804 | XP_011529132.1 | |
| XM_011530831.2 | 20 | 3796 | XP_011529133.1 | |
| XM_017029886.1 | XP_016885375.1 | |||
| XM_017029887.1 | XP_016885376.1 | |||
| XM_017029888.1 | XP_016885377.1 | |||
| XM_017029889.1 | XP_016885378.1 | |||
| AK304732.1 | ||||
| BC054499.2 | 26 | 4726 | AAH54499.1 | |
| EF613277.1 | ||||
| L25270.1 | 26 | 4973 | AAA61302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map