Assay Details
Target Gene Details
Entrez Gene ID: | 85369 |
Gene Name: | striatin interacting protein 1 |
Gene Aliases: |
FAM40A, FAR11A |
Location: |
Chr.1:110031577-110054641 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRIP1 | NM_001270768.1 | 10 | 1005 | NP_001257697.1 |
| NM_033088.3 | 10 | 1158 | NP_149079.2 | |
| NR_073071.1 | 11 | 1272 | ||
| XM_006710995.2 | 3 | 261 | XP_006711058.1 | |
| XM_017002629.1 | 10 | 1157 | XP_016858118.1 | |
| XM_017002630.1 | 10 | 1157 | XP_016858119.1 | |
| AB051548.2 | 10 | 1123 | BAB21852.2 | |
| AK027649.1 | 10 | 1005 | BAB55265.1 | |
| AK125054.1 | 3 | 927 | BAC86034.1 | |
| AK291059.1 | 10 | 1132 | ||
| AL834196.1 | 11 | 1241 | CAH56368.1 | |
| BC019919.1 | AAH19919.1 | |||
| BC094786.1 | 10 | 1139 | AAH94786.1 | |
| BC119814.1 | 10 | 1140 | ||
| BC121793.1 | 10 | 1140 |
More Information
Set Membership: |
|

Genomic Map