Assay Details
Target Gene Details
Entrez Gene ID: | 5097 |
Gene Name: | protocadherin 1 |
Gene Aliases: |
PC42, PCDH42 |
Location: |
Chr.5:141852319-141878410 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH1 | NM_032420.3 | 5 | 4418 | NP_115796.2 |
| XM_005268452.2 | 5 | 6748 | XP_005268509.1 | |
| XM_005268455.2 | 4 | 3441 | XP_005268512.1 | |
| XM_017009517.1 | 4 | 4375 | XP_016865006.1 | |
| AK075233.1 | 4 | 3787 | ||
| BM680840.1 | 1 | 393 | ||
| BU743106.1 | 1 | 393 | ||
| L11369.1 | AAA36418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
| nsv509093 | Chr.5:141806786 - 141884647 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map