Assay Details
Target Gene Details
Entrez Gene ID: | 7775 |
Gene Name: | zinc finger protein 232 |
Gene Aliases: |
ZSCAN11 |
Location: |
Chr.17:5105734-5123116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF232 | NM_001320952.1 | NP_001307881.1 | ||
| NM_001320953.1 | NP_001307882.1 | |||
| NM_001320954.1 | NP_001307883.1 | |||
| NM_001320955.1 | NP_001307884.1 | |||
| NM_014519.3 | NP_055334.2 | |||
| XM_011524006.2 | XP_011522308.1 | |||
| XM_017025021.1 | XP_016880510.1 | |||
| XM_017025022.1 | XP_016880511.1 | |||
| XM_017025023.1 | XP_016880512.1 | |||
| AK057506.1 | 2 | 629 | ||
| AK225622.1 | ||||
| AK297835.1 | 2 | 541 | ||
| AK298190.1 | 3 | 688 | ||
| AK316369.1 | 4 | 1300 | ||
| BC016039.1 | AAH16039.2 | |||
| Y15067.1 | CAB95963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map