Assay Details
Target Gene Details
Entrez Gene ID: | 60677 |
Gene Name: | CUGBP, Elav-like family member 6 |
Gene Aliases: |
BRUNOL6 |
Location: |
Chr.15:72284727-72320184 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF6 | NM_001172684.1 | NP_001166155.1 | ||
| NM_001172685.1 | NP_001166156.1 | |||
| NM_052840.4 | NP_443072.3 | |||
| AF401233.1 | AAK95615.1 | |||
| AF425606.1 | AAL34513.1 | |||
| AK124858.1 | ||||
| AK131098.1 | BAC85148.1 | |||
| AK294382.1 | ||||
| AK296004.1 | ||||
| BC030835.1 | AAH30835.1 | |||
| BC033838.2 | AAH33838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1601 | Chr.15:72257544 - 72292835 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map