Assay Details
Target Gene Details
Entrez Gene ID: | 221061 |
Gene Name: | family with sequence similarity 171 member A1 |
Gene Aliases: |
C10orf38 |
Location: |
Chr.10:15211643-15374532 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM171A1 | NM_001010924.1 | NP_001010924.1 | ||
| XM_011519378.2 | XP_011517680.1 | |||
| XM_017015904.1 | XP_016871393.1 | |||
| XM_017015905.1 | XP_016871394.1 | |||
| AL050367.1 | CAB43675.1 | |||
| AY683003.1 | AAV85904.1 | |||
| BC109302.1 | AAI09303.1 | |||
| BC109303.1 | AAI09304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509344 | Chr.10:15171456 - 15289792 on Build GRCh38 | Insertion |
|

Genomic Map