Assay Details
Target Gene Details
Entrez Gene ID: | 167127 |
Gene Name: | UDP glycosyltransferase family 3 member A2 |
Gene Aliases: |
- |
Location: |
Chr.5:36035017-36071358 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT3A2 | NM_001168316.1 | 4 | 949 | NP_001161788.1 |
| NM_174914.3 | 5 | 1051 | NP_777574.2 | |
| NR_031764.1 | 4 | 519 | ||
| XM_011513988.2 | 6 | 1093 | XP_011512290.1 | |
| XM_017009150.1 | 5 | 1102 | XP_016864639.1 | |
| AK075383.1 | 5 | 1012 | BAC11583.1 | |
| AK300751.1 | 4 | 910 | ||
| AK303154.1 | 4 | 480 | ||
| AY358416.1 | 5 | 986 | AAQ88782.1 | |
| BC103924.1 | 5 | 1000 | AAI03925.1 | |
| BC103925.1 | 5 | 1000 | AAI03926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map