Assay Details
Target Gene Details
Entrez Gene ID: | 54617 |
Gene Name: | INO80 complex subunit |
Gene Aliases: |
INO80A, INOC1, hINO80 |
Location: |
Chr.15:40978880-41117081 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80 | NM_017553.2 | NP_060023.1 | ||
| NR_104038.1 | ||||
| XM_011521686.2 | XP_011519988.1 | |||
| AB033085.1 | ||||
| AK002176.1 | BAA92122.1 | |||
| AK026050.1 | ||||
| AK057582.1 | ||||
| AK290371.1 | ||||
| BC146785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053462 | Chr.15:40977869 - 41103896 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map