Assay Details
Target Gene Details
Entrez Gene ID: | 57461 |
Gene Name: | ISY1 splicing factor homolog |
Gene Aliases: |
FSAP33 |
Location: |
Chr.3:129127416-129161230 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISY1 | NM_001199469.1 | NP_001186398.1 | ||
| NM_020701.3 | NP_065752.1 | |||
| BC004442.1 | AAH04442.1 | |||
| BC007409.2 | AAH07409.1 | |||
| BC019849.1 | AAH19849.1 |
Target Gene Details
Entrez Gene ID: | 100534599 |
Gene Name: | ISY1-RAB43 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:129087569-129161230 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISY1-RAB43 | NM_001204890.1 | NP_001191819.1 | ||
| AB032986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591650 | Chr.3:128986168 - 129177231 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map