Assay Details
Target Gene Details
Entrez Gene ID: | 93589 |
Gene Name: | calcium voltage-gated channel auxiliary subunit alpha2delta 4 |
Gene Aliases: |
RCD4 |
Location: |
Chr.12:1791957-1922606 on Build GRCh38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA2D4 | NM_172364.4 | 38 | 3827 | NP_758952.4 |
| AF516695.1 | 38 | 3959 | AAN06672.1 | |
| AK301234.1 | 11 | 1068 | ||
| AL137658.1 | 15 | 1366 | ||
| BC048288.1 | 10 | 933 | AAH48288.1 | |
| BC150186.1 | 37 | 3750 | ||
| BX537436.1 | 36 | 4427 | CAD97678.1 | |
| BX537437.1 | 38 | 3828 | CAD97679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832305 | Chr.12:1660686 - 1826888 on Build GRCh38 | Gain |
|
| nsv556996 | Chr.12:1782206 - 1825754 on Build GRCh38 | Gain |
|
| nsv832306 | Chr.12:1738560 - 1892142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map