Assay Details
Target Gene Details
Entrez Gene ID: | 51362 |
Gene Name: | cell division cycle 40 |
Gene Aliases: |
EHB3, PRP17, PRPF17 |
Location: |
Chr.6:110180421-110232220 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC40 | NM_015891.2 | NP_056975.1 | ||
| AB208903.1 | BAD92140.1 | |||
| AF015044.1 | AAD01552.1 | |||
| AF038392.1 | AAC39730.1 | |||
| AF061241.1 | AAC25166.1 | |||
| AK222721.1 | BAD96441.1 | |||
| AK295055.1 | ||||
| AK314601.1 | ||||
| BC111061.1 | 2 | 93 | AAI11062.1 | |
| BC117145.1 | ||||
| BC126114.1 |
Target Gene Details
Entrez Gene ID: | 8936 |
Gene Name: | WAS protein family member 1 |
Gene Aliases: |
SCAR1, WAVE, WAVE1 |
Location: |
Chr.6:110099817-110189874 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WASF1 | XM_011536233.1 | XP_011534535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759462 | Chr.6:110068723 - 110183477 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map