Assay Details
Target Gene Details
Entrez Gene ID: | 285126 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C5 gamma |
Gene Aliases: |
CSP-gamma |
Location: |
Chr.2:27275421-27281430 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC5G | NM_001303127.1 | 7 | 1556 | NP_001290056.1 |
| NM_001303128.1 | 6 | 1356 | NP_001290057.1 | |
| NM_173650.2 | 7 | 1618 | NP_775921.1 | |
| NR_130126.1 | 4 | 976 | ||
| XM_006711995.3 | 7 | 1530 | XP_006712058.1 | |
| XM_017003872.1 | 7 | 1568 | XP_016859361.1 | |
| XM_017003873.1 | 7 | 1557 | XP_016859362.1 | |
| XM_017003874.1 | 6 | 1332 | XP_016859363.1 | |
| AI183575.1 | 1 | 393 | ||
| AK097736.1 | 7 | 1618 | BAC05155.1 | |
| BC035194.2 | 7 | 1515 | AAH35194.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map