Assay Details
Target Gene Details
Entrez Gene ID: | 1950 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
HOMG4, URG |
Location: |
Chr.4:109912883-110013079 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGF | NM_001178130.2 | 1 | 295 | NP_001171601.1 |
| NM_001178131.2 | 1 | 295 | NP_001171602.1 | |
| NM_001963.5 | 1 | 295 | NP_001954.2 | |
| XM_005262796.2 | 1 | 296 | XP_005262853.1 | |
| XM_017007845.1 | 1 | 296 | XP_016863334.1 | |
| XM_017007846.1 | 1 | 296 | XP_016863335.1 | |
| XM_017007847.1 | 1 | 296 | XP_016863336.1 | |
| XM_017007848.1 | 1 | 296 | XP_016863337.1 | |
| XM_017007849.1 | 1 | 296 | XP_016863338.1 | |
| XM_017007850.1 | 1 | 296 | XP_016863339.1 | |
| XM_017007851.1 | 1 | 296 | XP_016863340.1 | |
| XM_017007852.1 | 1 | 296 | XP_016863341.1 | |
| XM_017007853.1 | 1 | 296 | XP_016863342.1 | |
| XM_017007854.1 | 1 | 296 | XP_016863343.1 | |
| XM_017007855.1 | 1 | 296 | XP_016863344.1 | |
| AK299306.1 | 1 | 297 | ||
| DB227969.1 | 1 | 296 | ||
| X04571.1 | 1 | 279 | CAA28240.1 |
More Information
Set Membership: |
|

Genomic Map