Assay Details
Target Gene Details
Entrez Gene ID: | 84869 |
Gene Name: | carbonyl reductase 4 |
Gene Aliases: |
SDR45C1 |
Location: |
Chr.4:168894484-169010317 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBR4 | XM_005263315.2 | XP_005263372.1 | ||
| XM_017008782.1 | XP_016864271.1 |
Target Gene Details
Entrez Gene ID: | 23022 |
Gene Name: | palladin, cytoskeletal associated protein |
Gene Aliases: |
CGI-151, CGI151, MYN, PNCA1, SIH002 |
Location: |
Chr.4:168497039-168928457 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
More Information
Set Membership: |
|

Genomic Map