Assay Details
Target Gene Details
Entrez Gene ID: | 8915 |
Gene Name: | B-cell CLL/lymphoma 10 |
Gene Aliases: |
CARMEN, CIPER, CLAP, IMD37, c-E10, mE10 |
Location: |
Chr.1:85265776-85276904 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL10 | NM_001320715.1 | 1 | 105 | NP_001307644.1 |
| NM_003921.4 | 1 | 105 | NP_003912.1 | |
| AF082283.1 | AAC99767.1 | |||
| AJ006288.1 | CAA06955.1 | |||
| BI831624.1 | 1 | 105 |
Target Gene Details
Entrez Gene ID: | 646626 |
Gene Name: | uncharacterized LOC646626 |
Gene Aliases: |
- |
Location: |
Chr.1:85276358-85278088 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC646626 | NR_045484.1 | |||
| AK125723.1 | ||||
| BX104049.1 | ||||
| DB483614.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34165 | Chr.1:85076357 - 85380566 on Build GRCh38 | Loss |
|
| nsv830459 | Chr.1:85123902 - 85320628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map