Assay Details
Target Gene Details
Entrez Gene ID: | 10242 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 2 |
Gene Aliases: |
- |
Location: |
Chr.3:178536298-178844429 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB2 | NM_181361.2 | 1 | 188 | NP_852006.1 |
| XM_011512325.2 | 1 | 92 | XP_011510627.1 | |
| AK298291.1 | ||||
| BC017825.1 | 1 | 50 | AAH17825.1 | |
| BG701449.1 | 1 | 195 | ||
| DA532966.1 | 1 | 250 |
Target Gene Details
Entrez Gene ID: | 104797538 |
Gene Name: | KCNMB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:178525467-178860405 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB2-AS1 | NR_126560.1 | |||
| NR_126561.1 | ||||
| AF279780.1 | ||||
| AY769439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569136 | Chr.3:178522284 - 178539169 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map