Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM | NM_001005502.2 | 9 | 4173 | NP_001005502.1 |
| NM_001874.4 | 9 | 4205 | NP_001865.1 | |
| NM_198320.3 | 9 | 4191 | NP_938079.1 | |
| BQ706414.1 | 1 | 234 | ||
| CB993834.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74714610] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map