Assay Details
Target Gene Details
Entrez Gene ID: | 4038 |
Gene Name: | LDL receptor related protein 4 |
Gene Aliases: |
CLSS, CMS17, LRP-4, LRP10, MEGF7, SOST2 |
Location: |
Chr.11:46854715-46918622 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP4 | NM_002334.3 | 38 | 6914 | NP_002325.2 |
| XM_011520103.2 | 32 | 5984 | XP_011518405.1 | |
| XM_011520104.2 | 23 | 4451 | XP_011518406.1 | |
| AB011540.1 | 30 | 5684 | BAA32468.1 | |
| AB084910.1 | 38 | 6914 | BAD83615.1 | |
| AB231861.1 | 38 | 6808 | ||
| AK027175.1 | 2 | 1614 | ||
| BC037360.2 | 3 | 1517 | AAH37360.1 | |
| BC041048.2 | 3 | 1517 | AAH41048.1 | |
| BC136667.1 | 38 | 6835 | ||
| BC136668.1 | 38 | 6835 |
Target Gene Details
Entrez Gene ID: | 100507401 |
Gene Name: | LRP4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:46846411-46874416 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP4-AS1 | NR_038909.1 | |||
| DB443018.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79910473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map