Assay Details
Target Gene Details
Entrez Gene ID: | 115752 |
Gene Name: | DIS3 like exosome 3'-5' exoribonuclease |
Gene Aliases: |
DIS3L1 |
Location: |
Chr.15:66293257-66333898 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIS3L | NM_001143688.2 | 7 | 955 | NP_001137160.1 |
| NM_001323936.1 | 7 | 874 | NP_001310865.1 | |
| NM_001323937.1 | 6 | 776 | NP_001310866.1 | |
| NM_001323938.1 | 7 | 882 | NP_001310867.1 | |
| NM_001323939.1 | 8 | 1079 | NP_001310868.1 | |
| NM_001323940.1 | 7 | 955 | NP_001310869.1 | |
| NM_001323941.1 | 7 | 1350 | NP_001310870.1 | |
| NM_001323943.1 | 7 | 1149 | NP_001310872.1 | |
| NM_001323944.1 | 7 | 1149 | NP_001310873.1 | |
| NM_001323945.1 | 7 | 1423 | NP_001310874.1 | |
| NM_001323946.1 | 7 | 1076 | NP_001310875.1 | |
| NM_001323948.1 | 6 | 1020 | NP_001310877.1 | |
| NM_133375.4 | 7 | 905 | NP_588616.1 | |
| XM_005254146.3 | 7 | 1023 | XP_005254203.1 | |
| AB075835.1 | 6 | 612 | BAB85541.1 | |
| AK095407.1 | 7 | 934 | BAC04542.1 | |
| AK295392.1 | 7 | 782 | ||
| BC022089.2 | 7 | 840 | AAH22089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047588 | Chr.15:66227169 - 66394432 on Build GRCh38 | Loss |
|
| nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map