Assay Details
Target Gene Details
Entrez Gene ID: | 90993 |
Gene Name: | cAMP responsive element binding protein 3 like 1 |
Gene Aliases: |
OASIS |
Location: |
Chr.11:46277638-46321422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L1 | NM_052854.3 | 1 | 493 | NP_443086.1 |
| XM_006718380.3 | 1 | 470 | XP_006718443.1 | |
| AB063321.1 | 1 | 114 | BAC01278.1 | |
| AK314072.1 | 1 | 471 | ||
| BC014097.1 | 1 | 136 | AAH14097.1 | |
| BC015781.1 | 1 | 122 | AAH15781.1 | |
| BQ953032.1 | 1 | 453 | ||
| CF131922.1 | 1 | 493 | ||
| DA693632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626183 | Chr.11:46227941 - 46507334 on Build GRCh38 | Gain |
|
| dgv197e214 | Chr.11:46176556 - 46312868 on Build GRCh38 | Gain |
|
| nsv554201 | Chr.11:46260284 - 46401136 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map