Assay Details
Target Gene Details
Entrez Gene ID: | 399665 |
Gene Name: | family with sequence similarity 102 member A |
Gene Aliases: |
C9orf132, EEIG1, SYM-3A, bA203J24.7 |
Location: |
Chr.9:127940582-127980533 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM102A | NM_001035254.2 | 11 | 3941 | NP_001030331.1 |
| NM_203305.2 | 8 | 3423 | NP_976050.1 | |
| AL049365.1 | 1 | 2322 | CAH10739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615381 | Chr.9:127906678 - 127954482 on Build GRCh38 | Loss |
|
| nsv1054891 | Chr.9:127939274 - 128102397 on Build GRCh38 | Gain |
|
| nsv831727 | Chr.9:127929082 - 128072682 on Build GRCh38 | Loss |
|
| nsv6722 | Chr.9:127912157 - 127945988 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111686473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map