Assay Details
Target Gene Details
Entrez Gene ID: | 27230 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
RAMP4 |
Location: |
Chr.3:150541993-150603177 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERP1 | NM_014445.3 | 3 | 2213 | NP_055260.1 |
| XM_011512698.2 | 4 | 2878 | XP_011511000.1 | |
| AK054644.1 | 1 | 1039 | ||
| AK090933.1 | 4 | 2025 | ||
| AK093929.1 | 4 | 2034 | ||
| AK098135.1 | 4 | 1929 | ||
| AK125413.1 | 4 | 2979 | ||
| AL136807.1 | 3 | 2022 | CAB66741.1 | |
| BC108314.1 | 3 | 2061 | AAI08315.1 |
More Information
Set Membership: |
|

Genomic Map